Fumarsäuremonomethylester, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(E)-4-methoxy-4-oxobut-2-enoic acid, Monomethyl Fumarate, Fumaric acid monomethyl ester, Methyl Hydrogen Fumarate
M-Clarity™ Qualitätsstufe = MQ100
651419-10G
651419-1G
SIAL651419-10GEA
133
EUR
SIAL651419-10G
SIAL651419-1G
Fumarsäuremonomethylester, Sigma-Aldrich®
Fumarsäuremonomethylester
Formel:
C₅H₆O₄ Molecular Weight: 130,1 g/mol |
MDL:
MFCD00063174 CAS-Nummer: 2756-87-8 EINECS: 220-412-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...