4-Methoxyphenylhydraziniumchlorid

Lieferant: ABCR

Synonyme: 4-Methoxyphenylhydrazine hydrochloride

AB110470.0050 AB110470.0250
ABCRAB110470.0050EA 0 EUR
ABCRAB110470.0050 ABCRAB110470.0250
4-Methoxyphenylhydraziniumchlorid
4-Methoxyphenylhydraziniumchlorid
Formel: C₇H₁₁ClN₂O
Molecular Weight: 174,63 g/mol
MDL: MFCD00012945
CAS-Nummer: 19501-58-7
EINECS: 243-115-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR