4-Methoxyphenylhydraziniumchlorid 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Methoxyphenylhydrazine hydrochloride

166490100 166490500
ACRO166490100EA 104 EUR
ACRO166490100 ACRO166490500
4-Methoxyphenylhydraziniumchlorid 98%
4-Methoxyphenylhydraziniumchlorid
Formel: C₇H₁₁ClN₂O
Molecular Weight: 174,63 g/mol
Schmelzpunkt: 153…154 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012945
CAS-Nummer: 19501-58-7
EINECS: 243-115-3
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance Light pink to grey to purple Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 155 to 163 °C
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR