4-Butoxy-2-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-N-Butoxy-2-methylphenylboronic acid

BB-8027-1G BB-8027-250M
COBBBB-8027-1GEA 88.6 EUR
COBBBB-8027-1G COBBBB-8027-250M
4-Butoxy-2-methylphenylboronsäure
4-Butoxy-2-methylphenylboronsäure
Formel: C₁₁H₁₇BO₃
Molecular Weight: 208,06 g/mol
MDL: MFCD05664325
CAS-Nummer: 845551-43-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR