2-Brom-5-(trifluormethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Bromo-5-(trifluoromethyl)phenyl)boronic acid, 2-Bromo-5-trifluoromethylphenylboronic acid

BB-3811-5G BB-3811-1G
COBBBB-3811-5GEA 449 EUR
COBBBB-3811-5G COBBBB-3811-1G
2-Brom-5-(trifluormethyl)phenylboronsäure
2-Brom-5-(trifluormethyl)phenylboronsäure
Formel: C₇H₅BBrF₃O₂
Molecular Weight: 268.82 g/mol
MDL: MFCD09258742
CAS-Nummer: 957034-38-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR