Silber-p-toluolsulfonat ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Toluene 4-sulphonic acid silver salt
L00307.06
L00307.14
L00307.06EA
44.3
EUR
L00307.06
L00307.14
Silber-p-toluolsulfonat ≥98%
Silber-p-toluolsulfonat
Purity: 98+%
CAS No.: 16836-95-6
Molecular Formula: C7H7AgO3S
Formula Weight: 279.07
Odour: Odourless
UN No.: 3077
MDL No.: MFCD00007511
Formel:
C₇H₇AgO₃S Molecular Weight: 279,06 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007511 CAS-Nummer: 16836-95-6 EINECS: 240-859-0 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...