Chlordimethyl(pentafluorphenyl)silan (Flophemesylchlorid)

Lieferant: CHEMPUR SILANE

Synonyme: (Chloro)(perfluorophenyl)dimethylsilane

SILS08950.1 SILS08950.5
SILNSILS08950.1EA 0 EUR
SILNSILS08950.1 SILNSILS08950.5
Chlordimethyl(pentafluorphenyl)silan (Flophemesylchlorid)
Chlordimethyl(pentafluorphenyl)silan (Flophemesylchlorid)
Formel: C₈H₆ClF₅Si
Molecular Weight: 260,67 g/mol
MDL: MFCD00000500
CAS-Nummer: 20082-71-7
EINECS: 243-506-9
UN: 2987
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR