Dimethylphenylsilan ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Dimethyl(phenyl)silane

L04558.09 L04558.18
L04558.09EA 49 EUR
L04558.09 L04558.18
Dimethylphenylsilan ≥97%
Dimethylphenylsilan

Purity: 97% CAS No.: 766-77-8 Molecular Formula: C8H12Si Formula Weight: 136.27 Flash Point: 35°(95°F) Refractive Index: 1.4970 Melting Point: -124° Boiling Point: 156-157° Beilstein Reference: 2204906 UN No.: 1993 MDL No.: MFCD00008257

Formel: C₈H₁₂Si
Molecular Weight: 136,27 g/mol
Siedepunkt: 156…157 °C
Schmelzpunkt: –124 °C
Dichte: 0,879
Flammpunkt: 35 °C (95 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008257
CAS-Nummer: 766-77-8
EINECS: 212-170-5
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR