Dimethylphenylsilan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Dimethyl(phenyl)silane

M-Clarity™ Qualitätsstufe = MQ100

235016-25G 235016-5G
SIAL235016-25GEA 134 EUR
SIAL235016-25G SIAL235016-5G
Dimethylphenylsilan, Sigma-Aldrich®
Dimethylphenylsilan
Formel: C₈H₁₂Si
Molecular Weight: 136,27 g/mol
MDL: MFCD00008257
CAS-Nummer: 766-77-8
EINECS: 212-170-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR