Dimethylphenylsilan

Lieferant: CHEMPUR SILANE

Synonyme: Dimethyl(phenyl)silane

SILS13425.10 SILS13425.25
SILNSILS13425.10EA 0 EUR
SILNSILS13425.10 SILNSILS13425.25
Dimethylphenylsilan
Dimethylphenylsilan
Formel: C₈H₁₂Si
Molecular Weight: 136,27 g/mol
MDL: MFCD00008257
CAS-Nummer: 766-77-8
EINECS: 212-170-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR