Bis(trimethylsilylmethyl)sulfid
Lieferant: CHEMPUR SILANE
Synonyme:
Bis(trimethylsilylmethyl) sulphide
SILS02775.1
SILS02775.5
SILNSILS02775.1EA
0
EUR
SILNSILS02775.1
SILNSILS02775.5
Bis(trimethylsilylmethyl)sulfid
Bis(trimethylsilylmethyl)sulfid
Formel:
C₈H₂₂SSi₂ Molecular Weight: 206.5 g/mol |
MDL:
MFCD00039797 CAS-Nummer: 4712-51-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...