Bis(4-hydroxyphenyl)sulfid ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
4,4'-Thiodiphenol
L04308.22
L04308.36
L04308.22EA
48.5
EUR
L04308.22
L04308.36
Bis(4-hydroxyphenyl)sulfid ≥98%
Bis(4-hydroxyphenyl)sulfid
Purity: 98+%
CAS No.: 2664-63-3
Molecular Formula: C12H10O2S
Formula Weight: 218.27
Melting Point: 151-155°
Beilstein Reference: 2050739
UN No.: 3261
MDL No.: MFCD00002349
Formel:
C₁₂H₁₀O₂S Schmelzpunkt: 151…155 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002349 CAS-Nummer: 2664-63-3 EINECS: 220-197-9 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...