N-Phenyl-2-naphthylamin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

178055-25G 178055-100G
SIAL178055-25GEA 37.8 EUR
SIAL178055-25G SIAL178055-100G
N-Phenyl-2-naphthylamin, Sigma-Aldrich®
N-Phenyl-2-naphthylamin
Formel: C₁₆H₁₃N
Molecular Weight: 219,29 g/mol
MDL: MFCD00004052
CAS-Nummer: 135-88-6
EINECS: 205-223-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR