2-Chlor-6-hydrazinopyridin
Lieferant: BIONET RESEARCH
Synonyme:
(6-Chloro-2-pyridyl)hydrazine
3B-055.500
3B-055.1
3B-055.5
3B-055.10
BIOT3B-055.500EA
0
EUR
BIOT3B-055.500
BIOT3B-055.1
BIOT3B-055.5
BIOT3B-055.10
2-Chlor-6-hydrazinopyridin
2-Chlor-6-hydrazinopyridin
Formel:
C₅H₆ClN₃ Molecular Weight: 143.58 g/mol |
MDL:
MFCD00214724 CAS-Nummer: 5193-03-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...