2-Chlor-6-hydrazinopyridin
Lieferant: COMBI-BLOCKS
Synonyme:
(6-Chloro-2-pyridyl)hydrazine
PY-7312-25G
PY-7312-5G
COBBPY-7312-25GEA
93.4
EUR
COBBPY-7312-25G
COBBPY-7312-5G
2-Chlor-6-hydrazinopyridin
2-Chlor-6-hydrazinopyridin
Formel:
C₅H₆ClN₃ Molecular Weight: 143.58 g/mol |
MDL:
MFCD00214724 CAS-Nummer: 5193-03-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...