(±)-1,2,3,4-Tetrahydro-1-naphthalinol

Lieferant: CHEMPUR

Synonyme: ac-alpha-Tetralol, 1,2,3,4-Tetrahydronaphthalen-1-ol, 1,2,3,4-Tetrahydro-1-naphthol

FM00150.250 FM00150.1
CHMPFM00150.250EA 0 EUR
CHMPFM00150.250 MAYBFM00150.1
(±)-1,2,3,4-Tetrahydro-1-naphthalinol
(±)-1,2,3,4-Tetrahydro-1-naphthol
Formel: C₁₀H₁₂O
Molecular Weight: 148,2 g/mol
MDL: MFCD00001739
CAS-Nummer: 529-33-9
EINECS: 208-459-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR