(R)-(+)-4-Isopropyl-2-oxazolidinon 98%

Lieferant: Thermo Fisher Scientific

Synonyme: (R)-4-Isopropyloxazolidin-2-one, (4R)-(+)-4-Isopropyl-2-oxazolidinone

297850010 297850050 297850250
ACRO297850010EA 58.6 EUR
ACRO297850010 ACRO297850050 ACRO297850250
(R)-(+)-4-Isopropyl-2-oxazolidinon 98%
(R)-(+)-4-Isopropyl-2-oxazolidinon
Formel: C₆H₁₁NO₂
Molecular Weight: 129.16 g/mol
Schmelzpunkt: 70…74 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00075172
CAS-Nummer: 95530-58-8

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 70 to 74 °C
GC ≥97.5 %
Specific optical rotation +16° to +20° (20 °C, 589 nm) (c=1, ethanol)

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR