(S)-(-)-4-Isopropyl-2-oxazolidinon 98%

Lieferant: Thermo Scientific

Synonyme: (4S)-(-)-Isopropyl-2-oxazolidinone, (4S)-(-)-4-Isopropyl-2-oxazolidinone, (S)-4-Isopropyl-2-oxazolidinone, (4S)-2-Oxo-4-(prop-2-yl)-1,3-oxazolidine, (4S)-4-(Prop-2-yl)-1,3-oxazolidin-2-one

297860010 297860050 297860250
ACRO297860010EA 30.6 EUR
ACRO297860010 ACRO297860050 ACRO297860250
(S)-(-)-4-Isopropyl-2-oxazolidinon 98%
(S)-(-)-4-Isopropyl-2-oxazolidinon
Formel: C₆H₁₁NO₂
Molecular Weight: 129,16 g/mol
Schmelzpunkt: 70…73 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010847
CAS-Nummer: 17016-83-0

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 70 to 74 °C
GC ≥97.5 %
Specific optical rotation -16° to -20° (20 °C, 589 nm) (c=1, ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR