(S)-(-)-4-Isopropyl-2-oxazolidinon ≥98%
Lieferant: Thermo Scientific
Synonyme:
(4S)-(-)-Isopropyl-2-oxazolidinone, (4S)-(-)-4-Isopropyl-2-oxazolidinone, (S)-4-Isopropyl-2-oxazolidinone, (4S)-2-Oxo-4-(prop-2-yl)-1,3-oxazolidine, (4S)-4-(Prop-2-yl)-1,3-oxazolidin-2-one
A14029.03
A14029.06
A14029.03EA
56.4
EUR
A14029.03
A14029.06
(S)-(-)-4-Isopropyl-2-oxazolidinon ≥98%
(S)-(-)-4-Isopropyl-2-oxazolidinon
Purity: 98%
CAS No.: 17016-83-0
Molecular Formula: C6H11NO2
Formula Weight: 129.16
Melting Point: 70-74°
Beilstein Reference: 3588198
Formel:
C₆H₁₁NO₂ Molecular Weight: 129,16 g/mol Schmelzpunkt: 70…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010847 CAS-Nummer: 17016-83-0 |