(R)-(+)-4-Benzyl-2-oxazolidinon ≥99%

Lieferant: Thermo Scientific

Synonyme: (R)-4-Benzyl-2-oxazolidinone

A16770.03 A16770.06 A16770.14
A16770.03EA 40.8 EUR
A16770.03 A16770.06 A16770.14
(R)-(+)-4-Benzyl-2-oxazolidinon ≥99%
(R)-(+)-4-Benzyl-2-oxazolidinon

Purity: 99% CAS No.: 102029-44-7 Molecular Formula: C10H11NO2 Formula Weight: 177.20 Melting Point: 86-89° Beilstein Reference: 4782551

Formel: C₁₀H₁₁NO₂
Molecular Weight: 177,2 g/mol
Schmelzpunkt: 86…89 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010846
CAS-Nummer: 102029-44-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR