(S)-(-)-4-Benzyl-2-oxazolidinon 99%, >99% ee

Lieferant: Thermo Scientific
Achtung

Synonyme: (S)-4-Benzyl-2-oxazolidinone, (S)-4-Benzyloxazolidin-2-one

402160025 402160250
ACRO402160025EA 60.1 EUR
ACRO402160025 ACRO402160250
(S)-(-)-4-Benzyl-2-oxazolidinon 99%, >99% ee
(S)-(-)-4-Benzyl-2-oxazolidinon
Formel: C₁₀H₁₁NO₂
Molecular Weight: 177,2 g/mol
Schmelzpunkt: 87…90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064496
CAS-Nummer: 90719-32-7

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 87 to 90 °C
HPLC ≥98.5 %
Enantiomeric excess ≥99 %
Specific optical rotation -60° to -64° (20 °C, 589 nm) (c=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR