(S)-(-)-4-Benzyl-2-oxazolidinon ≥99%

Lieferant: Thermo Scientific

Synonyme: (S)-4-Benzyl-2-oxazolidinone, (S)-4-Benzyloxazolidin-2-one

A18236.06 A18236.14
A18236.06EA 108 EUR
A18236.06 A18236.14
(S)-(-)-4-Benzyl-2-oxazolidinon ≥99%
(S)-(-)-4-Benzyl-2-oxazolidinon

Purity: 99% CAS No.: 90719-32-7 Molecular Formula: C10H11NO2 Formula Weight: 177.20 Melting Point: 87-89° Beilstein Reference: 3649667

Formel: C₁₀H₁₁NO₂
Molecular Weight: 177,2 g/mol
Schmelzpunkt: 87…89 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064496
CAS-Nummer: 90719-32-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR