(S)-(-)-4-Benzyl-2-oxazolidinon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-4-Benzyl-2-oxazolidinone, (S)-4-Benzyloxazolidin-2-one

M-Clarity™ Qualitätsstufe = MQ200

294640-25G 294640-5G
SIAL294640-25GEA 485 EUR
SIAL294640-25G SIAL294640-5G
(S)-(-)-4-Benzyl-2-oxazolidinon, Sigma-Aldrich®
(S)-(-)-4-Benzyl-2-oxazolidinon
Formel: C₁₀H₁₁NO₂
Molecular Weight: 177,2 g/mol
MDL: MFCD00064496
CAS-Nummer: 90719-32-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR