(R)-(-)-4-Phenyl-2-oxazolidinon ≥98%
Lieferant: Thermo Scientific
Synonyme:
(R)-4-Phenyloxazolidin-2-one
H27171.03
H27171.06
H27171.03EA
38.1
EUR
H27171.03
H27171.06
(R)-(-)-4-Phenyl-2-oxazolidinon ≥98%
(R)-(-)-4-Phenyl-2-oxazolidinon
Purity: 98%
CAS No.: 90319-52-1
Molecular Formula: C9H9NO2
Formula Weight: 163.18
Melting Point: 129-132°
Optical Rotation: -55° (c=2 in chloroform)
Formel:
C₉H₉NO₂ Molecular Weight: 163,18 g/mol Schmelzpunkt: 129…132 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00192393 CAS-Nummer: 90319-52-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...