(R)-(-)-4-Phenyl-2-oxazolidinon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (R)-4-Phenyloxazolidin-2-one

M-Clarity™ Qualitätsstufe = MQ100

402451-1G 402451-5G
SIAL402451-1GEA 74.5 EUR
SIAL402451-1G SIAL402451-5G
(R)-(-)-4-Phenyl-2-oxazolidinon, Sigma-Aldrich®
(R)-(-)-4-Phenyl-2-oxazolidinon
Formel: C₉H₉NO₂
Molecular Weight: 163,18 g/mol
MDL: MFCD00192393
CAS-Nummer: 90319-52-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR