(S)-(+)-4-Phenyl-2-oxazolidinon 99%

Lieferant: Thermo Scientific

Synonyme: (S)-4-Phenyloxazolidin-2-one

303850010 303850050
ACRO303850010EA 67.1 EUR
ACRO303850010 ACRO303850050
(S)-(+)-4-Phenyl-2-oxazolidinon 99%
(S)-(+)-4-Phenyl-2-oxazolidinon
Formel: C₉H₉NO₂
Molecular Weight: 163,18 g/mol
Schmelzpunkt: 130…133 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00043396
CAS-Nummer: 99395-88-7

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 130 to 133 °C
GC ≥98.5 %
Specific optical rotation +48° to +51° (20 °C, 589 nm) (c=2, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR