(R)-(+)-1,1'-Bi-2-naphthol ≥99%
Lieferant: Thermo Fisher Scientific
Gefahr
Synonyme: (R)-[1,1'-Binaphthalene]-2,2'-diol, (R)-BINOL, (R)-(+)-1,1'-Binaphthyl-2,2'-Diol
L08305.03 L08305.06 L08305.14
L08305.03EA 39.6 EUR
L08305.03 L08305.06 L08305.14
(R)-(+)-1,1'-Bi-2-naphthol ≥99%
(R)-(+)-1,1'-Bi-2-naphthol
Purity: 99% CAS No.: 18531-94-7 Molecular Formula: C20H14O2 Formula Weight: 286.33 Odour: Characteristic Melting Point: 206-211° Beilstein Reference: 3616837 Merck Reference: 14,1226 Optical Rotation: +34.5 (c=1 in THF) UN No.: 2811 MDL No.: MFCD00004068
Formel: C₂₀H₁₄O₂
Molecular Weight: 286,33 g/mol
Schmelzpunkt: 206…211 °C
Dichte: 1,301
Lagertemperatur: Raumtemperatur
MDL: MFCD00004068
CAS-Nummer: 18531-94-7
UN: 2811
ADR: 6.1,III
Merck Index: 14,1226