(S)-(-)-1,1'-Bi-2-naphthol ≥99%
Lieferant: Thermo Fisher Scientific
Gefahr
Synonyme: (S)-BINOL, (S)-(-)-1,1'-Binaphthyl-2,2'-Diol, (S)-[1,1'-Binaphthalene]-2,2'-diol
L08423.03 L08423.06 L08423.14
L08423.03EA 28.5 EUR
L08423.03 L08423.06 L08423.14
(S)-(-)-1,1'-Bi-2-naphthol ≥99%
(S)-(-)-1,1'-Bi-2-naphthol
Purity: 99% CAS No.: 18531-99-2 Molecular Formula: C20H14O2 Formula Weight: 286.33 Odour: Characteristic Melting Point: 206-211° Beilstein Reference: 3592999 Merck Reference: 14,1226 Optical Rotation: -34.5 (c=1 in THF) UN No.: 2811 MDL No.: MFCD00004068
Formel: C₂₀H₁₄O₂
Molecular Weight: 286,33 g/mol
Schmelzpunkt: 206…211 °C
Dichte: 1,301
Lagertemperatur: Raumtemperatur
MDL: MFCD19441803
CAS-Nummer: 18531-99-2
UN: 2811
ADR: 6.1,III
Merck Index: 14,1226