2-Amino-5-brom-3-nitropyridin

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Brom-3-nitropyridin-2-amin, 5-Brom-3-nitro-2-pyridylamin

BTB03307.1 BTB03307.10 BTB03307.50
MAYBBTB03307.1EA 0 EUR
MAYBBTB03307.1 MAYBBTB03307.10 MAYBBTB03307.50
2-Amino-5-brom-3-nitropyridin
2-Amino-5-brom-3-nitropyridin
Formel: C₅H₄BrN₃O₂
Molecular Weight: 218.01 g/mol
Schmelzpunkt: 215…216 °C
MDL: MFCD00047441
CAS-Nummer: 6945-68-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR