Furan-3-boronsäure

Lieferant: CHEMPUR

Synonyme: 3-Furanboronic acid, 3-Furanylboronic acid

CC02112.250 CC02112.1
CHMPCC02112.250EA 0 EUR
CHMPCC02112.250 MAYBCC02112.1
Furan-3-boronsäure
Furan-3-boronsäure
Formel: C₄H₅BO₃
Molecular Weight: 111,89 g/mol
MDL: MFCD01319007
CAS-Nummer: 55552-70-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR