6-Isochinolinol

Lieferant: COMBI-BLOCKS

Synonyme: 6-Hydroxyisoquinoline, Isoquinolin-6-ol

HC-6030-5G HC-6030-1G
COBBHC-6030-5GEA 0 EUR
COBBHC-6030-5G COBBHC-6030-1G
6-Isochinolinol
6-Isochinolinol
Formel: C₉H₇NO
MDL: MFCD04114860
CAS-Nummer: 7651-82-3
EINECS: 231-612-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR