2-Chlor-N-(2,6-dimethylphenyl)acetamid ≥99%

Lieferant: Thermo Scientific
Achtung

Synonyme: N-(2,6-Dimethylphenyl)chloroacetamide, 2-Chloro-2',6'-dimethylacetanilide, N-(2,6-Dichlorophenyl)-2-chloroacetamide

L02727.09 L02727.18
L02727.09EA 58.8 EUR
L02727.09 L02727.18
2-Chlor-N-(2,6-dimethylphenyl)acetamid ≥99%
2-Chlor-N-(2,6-dimethylphenyl)acetamid

Purity: 99% CAS No.: 1131-01-7 Molecular Formula: C10H12ClNO Formula Weight: 197.67 Melting Point: 146-150° Beilstein Reference: 781309

Formel: C₁₀H₁₂ClNO
Molecular Weight: 197,66 g/mol
Schmelzpunkt: 143…148 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00000926
CAS-Nummer: 1131-01-7
EINECS: 214-460-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR