Nitrofen ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2,4-Dichloro-1-(4-nitrophenoxy)benzene

A19977.14 A19977.06
A19977.14EA 208 EUR
A19977.14 A19977.06
Nitrofen ≥98%
Nitrofen

Purity: 98% CAS No.: 1836-75-5 Molecular Formula: C12H7Cl2NO3 Formula Weight: 284.10 Melting Point: 68-71° Beilstein Reference: 1887356 Merck Reference: 14,6598 UN No.: 3077 MDL No.: MFCD00128026

Formel: C₁₂H₇Cl₂NO₃
Molecular Weight: 284.1 g/mol
Schmelzpunkt: 68…71 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00128026
CAS-Nummer: 1836-75-5
EINECS: 217-406-0
ADR: 9 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR