Isochinolin-5-amin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Isoquinolin-5-amine, 5-Aminoisoquinoline

M-Clarity™ Qualitätsstufe = MQ100

136107-1G 136107-5G
SIAL136107-1GEA 34.5 EUR
SIAL136107-1G SIAL136107-5G
Isochinolin-5-amin, Sigma-Aldrich®
Isochinolin-5-amin
Formel: C₉H₈N₂
Molecular Weight: 144,18 g/mol
MDL: MFCD00006907
CAS-Nummer: 1125-60-6
EINECS: 214-408-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR