Isochinolin-8-amin

Lieferant: COMBI-BLOCKS

Synonyme: Isoquinolin-8-amine, 8-Aminoisoquinoline

HC-6001-1G HC-6001-5G
COBBHC-6001-1GEA 88.6 EUR
COBBHC-6001-1G COBBHC-6001-5G
Isochinolin-8-amin
Isochinolin-8-amin
Formel: C₉H₈N₂
Molecular Weight: 144.1700 g/mol
MDL: MFCD00179553
CAS-Nummer: 23687-27-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR