2-Fluor-4-methoxyphenylboronsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Fluoro-4-methoxybenzeneboronic acid

L19960.03 L19960.06
L19960.03EA 62.2 EUR
L19960.03 L19960.06
2-Fluor-4-methoxyphenylboronsäure ≥98%
2-Fluor-4-methoxyphenylboronsäure

Purity: 98% CAS No.: 162101-31-7 Molecular Formula: C7H8BFO3 Formula Weight: 169.95 Beilstein Reference: 9192698 MDL No.: MFCD04039323

Formel: C₇H₈BFO₃
Molecular Weight: 169.95 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04039323
CAS-Nummer: 162101-31-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR