2-Fluor-6-methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Fluoro-6-methoxybenzeneboronic acid

BB-2289-5G BB-2289-25G BB-2289-100G
COBBBB-2289-5GEA 49.7 EUR
COBBBB-2289-5G COBBBB-2289-25G COBBBB-2289-100G
2-Fluor-6-methoxyphenylboronsäure
2-Fluor-6-methoxyphenylboronsäure
Formel: C₇H₈BFO₃
Molecular Weight: 169,95 g/mol
MDL: MFCD02179483
CAS-Nummer: 78495-63-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR