3,5-Difluor-2-methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3,5-Difluoro-2-methoxyphenyl)boronic acid, 3,5-Difluoro-2-methoxybenzeneboronic acid

BB-8403-5G BB-8403-1G
COBBBB-8403-5GEA 486 EUR
COBBBB-8403-5G COBBBB-8403-1G
3,5-Difluor-2-methoxyphenylboronsäure
3,5-Difluor-2-methoxyphenylboronsäure
Formel: C₇H₇BF₂O₃
Molecular Weight: 187.94 g/mol
MDL: MFCD03095353
CAS-Nummer: 737000-76-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR