3-Fluor-4-methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-Fluoro-4-methoxyphenyl)boronic acid, 3-Fluoro-4-methoxybenzeneboronic acid

BB-2130-5G BB-2130-25G
COBBBB-2130-5GEA 75.4 EUR
COBBBB-2130-5G COBBBB-2130-25G
3-Fluor-4-methoxyphenylboronsäure
3-Fluor-4-methoxyphenylboronsäure
Formel: C₇H₈BFO₃
Molecular Weight: 169,95 g/mol
MDL: MFCD00807404
CAS-Nummer: 149507-26-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR