5-Fluor-2-methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-Fluoro-2-methoxybenzeneboronic acid

BB-2692-1G BB-2692-5G BB-2692-100G BB-2692-25G
COBBBB-2692-1GEA 0 EUR
COBBBB-2692-1G COBBBB-2692-5G COBBBB-2692-100G COBBBB-2692-25G
5-Fluor-2-methoxyphenylboronsäure
5-Fluor-2-methoxyphenylboronsäure
Formel: C₇H₈BFO₃
Molecular Weight: 169,95 g/mol
MDL: MFCD01863526
CAS-Nummer: 179897-94-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR