3-Methoxy-2,4,6-trifluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2,4,6-Trifluoro-3-methoxyphenyl)boronic acid

BB-8037-1G BB-8037-250M
COBBBB-8037-1GEA 269 EUR
COBBBB-8037-1G COBBBB-8037-250M
3-Methoxy-2,4,6-trifluorphenylboronsäure
3-Methoxy-2,4,6-trifluorphenylboronsäure
Formel: C₇H₆BF₃O₃
Molecular Weight: 205.927 g/mol
MDL: MFCD07369755
CAS-Nummer: 849062-08-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR