4-Fluor-2-methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Fluoro-2-methoxybenzeneboronic acid

BB-2686-5G BB-2686-25G BB-2686-1G
COBBBB-2686-5GEA 101 EUR
COBBBB-2686-5G COBBBB-2686-25G COBBBB-2686-1G
4-Fluor-2-methoxyphenylboronsäure
4-Fluor-2-methoxyphenylboronsäure
Formel: C₇H₈BFO₃
Molecular Weight: 169.95 g/mol
MDL: MFCD03788423
CAS-Nummer: 179899-07-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR