4-Fluor-2-methoxyphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Fluoro-2-methoxybenzeneboronic acid

M-Clarity™ Qualitätsstufe = MQ100

564494-1G 564494-5G
SIAL564494-1GEA 26 EUR
SIAL564494-1G SIAL564494-5G
4-Fluor-2-methoxyphenylboronsäure, Sigma-Aldrich®
4-Fluor-2-methoxyphenylboronsäure
Formel: C₇H₈BFO₃
Molecular Weight: 169,95 g/mol
MDL: MFCD03788423
CAS-Nummer: 179899-07-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR