4-Fluor-3-methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-Fluoro-3-methoxyphenyl)boronic acid, 4-Fluoro-3-methoxybenzeneboronic acid

BB-2809-5G BB-2809-1G
COBBBB-2809-5GEA 180 EUR
COBBBB-2809-5G COBBBB-2809-1G
4-Fluor-3-methoxyphenylboronsäure
4-Fluor-3-methoxyphenylboronsäure
Formel: C₇H₈BFO₃
Molecular Weight: 169,95 g/mol
MDL: MFCD08056358
CAS-Nummer: 854778-31-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR