2-(4-Nitrophenyl)ethanol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-Nitrophenethyl alcohol, p-Nitrophenethyl Alcohol
M-Clarity™ Qualitätsstufe = MQ100
183466-10G
183466-50G
SIAL183466-10GEA
35.8
EUR
SIAL183466-10G
SIAL183466-50G
2-(4-Nitrophenyl)ethanol, Sigma-Aldrich®
2-(4-Nitrophenyl)ethanol
Formel:
C₈H₉NO₃ Molecular Weight: 167.16 g/mol |
MDL:
MFCD00010202 CAS-Nummer: 100-27-6 EINECS: 202-835-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...