2-Chlor-N-(2,6-dimethylphenyl)acetamid
Lieferant: Thermo Scientific
Synonyme:
N-(2,6-Dimethylphenyl)chloroacetamide, 2-Chloro-2',6'-dimethylacetanilide, N-(2,6-Dichlorophenyl)-2-chloroacetamide
CD07038.250
CD07038.1
MAYBCD07038.250EA
0
EUR
MAYBCD07038.250
MAYBCD07038.1
2-Chlor-N-(2,6-dimethylphenyl)acetamid
2-Chlor-N-(2,6-dimethylphenyl)acetamid
Formel:
C₁₀H₁₂ClNO Molecular Weight: 197,66 g/mol |
MDL:
MFCD00000926 CAS-Nummer: 1131-01-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...