N-(3,5-Dichlorphenyl)-2-chloracetamid

Lieferant: Thermo Scientific

Synonyme: N-(Chloroacetyl)-3,5-dichloroaniline

SPB05348.1 SPB05348.10 SPB05348.50
MAYBSPB05348.1EA 0 EUR
MAYBSPB05348.1 MAYBSPB05348.10 MAYBSPB05348.50
N-(3,5-Dichlorphenyl)-2-chloracetamid
N-(3,5-Dichlorphenyl)-2-chloracetamid
Formel: C₈H₆Cl₃NO
CAS-Nummer: 33560-48-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR