2',4'-Dichlor-6'-methylacetanilid

Lieferant: Thermo Scientific

TL00876.1 TL00876.10 TL00876.50
MAYBTL00876.1EA 0 EUR
MAYBTL00876.1 MAYBTL00876.10 MAYBTL00876.50
2',4'-Dichlor-6'-methylacetanilid
2',4'-Dichlor-6'-methylacetanilid
Formel: C₉H₉Cl₂NO
Molecular Weight: 218,08 g/mol
CAS-Nummer: 61655-97-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR