2,6-Difluornicotinsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2,6-Difluoropyridine-3-carboxylic acid

OT-8124-5G OT-8124-1G
COBBOT-8124-5GEA 63.4 EUR
COBBOT-8124-5G COBBOT-8124-1G
2,6-Difluornicotinsäure
2,6-Difluornicotinsäure
Formel: C₆H₃F₂NO₂
Molecular Weight: 159,0900 g/mol
MDL: MFCD08064047
CAS-Nummer: 171178-50-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR