Ethyl-3-hydroxybutyrat ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Ethyl 3-hydroxybutanoate, DL-Ethyl-3-hydroxybutyrate, Ethyl 3-hydroxybutyrate

B21521.14 B21521.22
B21521.14EA 36.7 EUR
B21521.14 B21521.22
Ethyl-3-hydroxybutyrat ≥98%
Ethyl-3-hydroxybutyrat

Purity: 98+% CAS No.: 5405-41-4 Molecular Formula: C6H12O3 Formula Weight: 132.16 Flash Point: 64°(148°F) Refractive Index: 1.4210 Boiling Point: 169-171° Beilstein Reference: 1446190 MDL No.: MFCD00004545

Formel: C₆H₁₂O₃
Molecular Weight: 132.16 g/mol
Siedepunkt: 169…171 °C
Dichte: 1.013
Flammpunkt: 64 °C (148 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004545
CAS-Nummer: 5405-41-4
EINECS: 226-456-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR